Learn More
Bel-Art™ Glove Box Economy Sleeve Gloves
Product Code. 11767385
Change view
Click to view available options
Size:
9
10
Unit Size:
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11767385 9 Pack of 2
11757385 10 Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11767385 Supplier Bel-Art™ Supplier No. H500250546
  • ONLINE EXCLUSIVE PRICE
    376.00€
    333.40€ / paquete de 2
    Ahorro 42.60€
    11% Descuento
Fecha estimada de envoi: 04-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
One piece, bonded neoprene protection against liquid or gas penetration
  • Single piece glove with sleeve
  • For glove boxes with 8 in. glove ports
  • Requires clamping rings (19-821-983) to hold gloves in place
  • Sold in pairs
Specifications
Color Black
Hand Specific
Latex Free No
Material Bonded Neoprene
Product Type Glove Box Gloves
Size 9