Learn More
behr Labor-Technik™ behrotest™ Extraction Device for Twisselmann Hot Extraction System
Product Code. 10245152
Change view
Click to view available options
Capacity (Metric):
6 x 100 mL
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
10245152 6 x 100 mL Each
1 options
This item is not returnable. View return policy
Product Code. 10245152 Supplier behr Labor-Technik™ Supplier No. B00218424
  • ONLINE EXCLUSIVE PRICE
    8481.93€
    4362.86€ / paquete de 1
    Ahorro 4119.07€
    48% Descuento
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Extractors for multi-samples.
  • Extraction according to Twisselmann method
  • Continuous hot extraction
  • Improves solubility and shortens extraction time
  • Energy individually adjustable for every sample position
  • Cooling water distribution strip ensures uniform cooling at sample positions
  • Practical condenser ledge to safely deposit condenser between hot extractions
  • Brackets for safely holding intermediate extraction pieces after removing sample container
Specifications
For Use With (Equipment) Twisselmann Hot Extraction System
Product Type Extraction Device
Capacity (Metric) 6 x 100 mL
Includes Brackets
For Use With (Application) Extraction of Solvents