missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brand™ Vasos de precipitados de polipropileno de perfil bajo con escala grabada
Click to view available options
Capacidad (métrico):
50 mL
100 mL
400 mL
2000 mL
3000 mL
Descripción
- With beaded rim and spout
- Autoclavable
Especificaciones
Capacidad (métrico) | 3000 mL |
Material | Polypropylene |
Incrementos | 250mL |
Graduated | Yes |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'