missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BDNF, Recombinant Protein, Invitrogen™
Recombinant Protein
Marca: Invitrogen RP10917
Detalles adicionales : Peso : 0.01000kg
Descripción
BDNF Recombinant Protein, RP-10917, from Invitrogen.
RP-10917 contains recombinant human BDNF protein suitable for use as a control in ELISA and Western blot applications. RP-10917 is reported to have the additional biological activity: ED50 as determined by the dose-dependent induction of acetylcholine esterase in rat basal forebrain primary septal culture is 50ng/ml. Upon reconstitution, store at 4ºC for 2-7 days or aliquot and store at -20ºC. For long term storage add either HSA or BSA to 0.1%. Avoid repeat freeze/thaw cycles. Reconstitute with 100 ug/ml of distilled water.Especificaciones
P23560 | |
0.02M citric acid with no preservative; pH 5 | |
Human BDNF | |
10μg | |
-20°C, Avoid Freeze/Thaw Cycles | |
Brain-derived Neurotrophic Factor | |
BDNF | |
Unconjugated | |
E. coli | |
Lyophilized | |
purified |
Control | |
627 | |
5 | |
E. coli | |
RUO | |
Human BDNF | |
Human | |
Hu | |
Recombinant | |
Recombinant |