Learn More
BD Vacutainer™ Tube Holder
Product Code. 10148594
Change view
Click to view available options
Size:
Standard
Unit Size:
Pack of 1000
Product Code. Size Unit Size
10148594 Standard Pack of 1000
1 options
This item is not returnable. View return policy
Product Code. 10148594 Supplier BD Supplier No. 364815
  • ONLINE EXCLUSIVE PRICE
    60.40€
    43.92€ / paquete de 1000
    Ahorro 16.48€
    27% Descuento
 Restricted Item
Fecha estimada de envoi: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Low-cost, single-use holder compliant with OSHA regulations
  • Compatible with the entire BD Vacutainer system of blood collection products
  • Standard size, clear
  • Non-sterile
  • Clear
  • Single use, non-stackable
  • Latex free
Compliance
Complies with OSHA Safety and Health Information Bulletin (SHIB) on the disposal of contaminated needles and blood tube holders used for phlebotomy
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Disposable Single-use
For Use With (Application) Reduces the hazards of contact with Blood borne pathogens compared to reusable holders
Latex Free Yes
Size Standard
Product Type Tube Holder
Sterility Non-sterile