missing translation for 'onlineSavingsMsg'
Learn More

BD Medios de cultivo deshidratados Difco™: agar de infusión de cerebro y corazón

Para aislamiento de Aspergillus brasiliensis (niger) 16404, Escherichia coli 25922, Staphylococcus aureus 25923, Streptococcus pneumoniae 6305, Streptococcus pyogenes 19615

Marca:  BD 241820

Detalles adicionales : Peso : 0.80000kg

 Ver más versiones de este producto

Código de producto. 11798862

  • 79.89€ / 100 g


Consúltenos : 902 239 303 
Añadir a la cesta

Descripción

Descripción

BHI Agar has proven to be effective in the cultivation of a wide variety of fastidious microorganisms, including many types of pathogens, fungi and yeasts. Also used with added antibiotics in isolating fungi. It derives its nutrients from the brain heart infusion, peptone and dextrose components.

  • The peptones and infusion are sources of organic nitrogen, carbon, sulfur, vitamins and trace substances
  • Dextrose is a carbohydrate source that microorganisms utilize by fermentative action
  • The medium is buffered through the use of disodium phosphate
  • To prepare solution, suspend 52g in 1L purified water
  • pH (5.2% solution, 25°C): 7.4 ± 0.2
  • Good cultural response with: Aspergillus brasiliensis (niger) 16404, Escherichia coli 25922, Staphylococcus aureus 25923, Streptococcus pneumoniae 6305, Streptococcus pyogenes 19615
Especificaciones

Especificaciones

ISO 9000, AOAC, BAM, CCAM, COMPF, EPA, MCM9, SMD, SMWW, USDA
Dehydrated
BHI Agar
100g
Brain heart infusion Agar
Beige
Calf brain infusion from 200g 7.7g/L, Beef heart infusion from 250g 9.8g/L, Proteose Peptone 10g/L, Dextrose 2g/L, Sodium Chloride 5g/L, Disodium Phosphate 2.5g/L, Agar 15g/L
Bottle
<30°C
Isolation of Aspergillus brasiliensis (niger) 16404, Escherichia coli 25922, Staphylococcus aureus 25923, Streptococcus pneumoniae 6305, Streptococcus pyogenes 19615
Videos
Documentos