missing translation for 'onlineSavingsMsg'
Learn More

BCS1L Mouse anti-Human, Mouse, Clone: 3G6H1, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 60212-1-IG-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817225

  • 399.00€ / 150 microlitros
Fecha estimada de envío: 17-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a homolog of the S. cerevisiae bcs1 protein which is involved in the assembly of complex III of the mitochondrial respiratory chain. The encoded protein does not contain a mitochondrial targeting sequence but experimental studies confirm that it is imported into mitochondria. Mutations in this gene are associated with mitochondrial complex III deficiency and the GRACILE syndrome. Five alternatively spliced transcripts encoding the same protein have been described.
Especificaciones

Especificaciones

BCS1L
Monoclonal
0.86 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9CZP5, Q9Y276
BCS1L
BCS1L Fusion Protein Ag18280
150 μL
Primary
Human, Mouse
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
3G6H1
Unconjugated
BCS1L
BCS1L, BJS, FLNMS, GRACILE, h BCS, h BCS1, Hs.6719, Mitochondrial chaperone BCS1, PTD
Mouse
Protein G
RUO
617, 66821
-20°C
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales