missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bcl-6 (Follicular Lymphoma Marker) Mouse, Clone: rBCL6/1527, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 604-MSM10-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Bcl-6 (Follicular Lymphoma Marker) | |
Nuclear | |
rBCL6/1527 | |
Unconjugated | |
Mouse | |
95kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
604 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human bcl-6 protein fragment (apprx. aa256-389) | |
Raji or Ramos cells. Tonsil or Hodgkin's lymphoma. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |