Learn More
Thermo Scientific Chemicals BCA Protein Assay Kit
Product Code. 15485549
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Product Code. Quantity Unit Size
15485549 1 Kit Each
1 options
This item is not returnable. View return policy
Product Code. 15485549 Supplier Thermo Scientific Chemicals Supplier No. J63283.QA
  • ONLINE EXCLUSIVE PRICE
    378.42€
    324.65€ / paquete de 1
    Ahorro 53.77€
    14% Descuento
 Restricted Item
En stock
Add to basket
Edge
This item is not returnable. View return policy
BCA Protein Assay Kit can be used to measure very dilute protein concentrations in very small sample volumes. Accurately measures protein concentrations from 0.5 to 30 ?g/mL in tube assays and 1 to 20 ?g/mL in 96- or 384-well plate assays. It is used for studying protein: protein interactions, measuring column fractions after affinity chromatography, estimating percent recovery of membrane proteins from cell extracts and high-throughput screening of fusion proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Description Clear
Product Type Protein Assay Kit
For Use With (Application) Contains two separate solutions for estimation of protein concentration.
Quantity 1 Kit
What BCA protein assay kits do you recommend for NanoDrop 2000/2000c Spectrophotometers (Cat. Nos. ND-2000, ND-2000C)?
NanoDrop 2000/2000c Spectrophotometers (Cat. Nos. ND-2000, ND-2000C) are compatible with most commercial BCA protein assay kits. We offer a number of BCA protein assay kits, including, but not limited to, the BCA Protein Assay Kit (Cat. No. J63283.QA), Micro BCA Protein Assay Kit (Cat. No. 23235, and Pierce BCA Protein Assay Kit (Cat. No 23225, 23227).
To identify, compare, and choose the best protein assay for your sample type and application, you can use our interactive Protein Assay Selection Guide.
What are the contents of the BCA Protein Assay Kit (Cat. No. J63283.QA)?
The BCA Protein Assay Kit contains 250 mL of Solution A and 5.5 mL of Solution B. The solution components are as follows:
  • Solution A: Bicinchonic acid and tartrate in an alkaline carbonate buffer
  • Solution B: 4% w/v cupric sulfate