Learn More
Fisherbrand™ Battery Acid Specialty Spill Kit
Product Code. 16563852 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Quantity: 1/Ea.
Product Code.
16563852 Quantity: 1/Ea.
1 options
Product Code. 16563852
  • ONLINE EXCLUSIVE PRICE
    209.52€
    160.48€ / 1 sets
    Ahorro 49.04€
    23% Descuento
 Restricted Item
Fecha estimada de envoi: 28-12-2026
Add to basket
Add to basket
Spill kit designed to pick up battery acid on both smooth and rough surfaces while remaining chemically inert.
  • Place near a battery swap station and around your facility
  • Allows you to be prepared should a spill occur
  • Designed to pick up battery acid on both smooth and rough surfaces
  • Chemically inert
Specifications
Type Chemical Sorbent
Includes 1 x BAG-DIS-YEL (834179), 1 x SPK-HAND-HAZ (198786), 2 x HAZ412 (813850), 10 x UN100-E (813842), 1 x SPK-BRIL (198784), 1 x SPK-SUITDIS (198790), 1 x SPC-ACID, 1 x Bucket, 1 x Scoop and Scraper, 1 x SF-JAR-CS (149265)
compliance-icons
Product Identifier
  • ACID NEUTRALIZER (shaker; pail)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: sodium carbonate