Learn More
Cytiva BAS Storage Phosphor Screens
Product Code. 15327457 Shop All Cytiva Products
Change view
Click to view available options
Detector Type:
Neutron
Unit Size:
Each
Product Code. Detector Type Unit Size
15327457 Neutron Each
1 options
This item is not returnable. View return policy
Product Code. 15327457 Supplier Cytiva Supplier No. 29017133
Fecha estimada de envoi: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For Neutron Detection, Size 20 x 40 cm
  • Requires approximately one-tenth the exposure time of traditional autoradiography film
  • Ten to one hundred times more sensitive than film, depending on isotope and sample type
  • Wide linear dynamic range (five orders of magnitude) enables quantitation and visualization of both weak and strong signals in a single exposure
  • Reusable screens do not require chemicals, darkroom, or other special treatment for use
  • Each step can be performed at the lab bench under normal lighting conditions at room temperature
  • Results are digitized using a storage phosphor imaging system (e.g., Typhoon, Storm, or PhosphorImager) and quantitated using ImageQuant software
TRUSTED_SUSTAINABILITY
Specifications
Description 20 x 40 cm
Detectors Neutron
Detector Type Neutron
For Use With (Application) Neutron Detection