Learn More
Labconco Balance Vibration Isolator
Product Code. 15522495 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Marble slab, Four isolator pad, Protective top cover
Unit Size:
Each
Product Code. Includes Unit Size
15522495 Marble slab, Four isolator pad, Protective top cover Each
1 options
This item is not returnable. View return policy
Product Code. 15522495 Supplier Labconco Supplier No. 5234600P
  • ONLINE EXCLUSIVE PRICE
    1310.00€
    1134.66€ / paquete de 1
    Ahorro 175.34€
    13% Descuento
Fecha estimada de envoi: 28-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Isolator minimizes vibration to sensitive balances
  • Includes 13.4 W x 20.4 D x 2.7 in. thick marble slab, four isolator pads, and type 304 Stainless Steel protective top cover
  • Included with XPert Weigh Boxes
  • Applicable Products:
    • Precise Basic Glove Boxes
    • Precise Controlled Atmosphere Glove Boxes
    • Precise Hepa-filtered Glove Boxes
    • Protector Combination Glove Boxes
    • Protector Controlled Atmosphere Glove Boxes
    • Protector Filtered Glove Boxes
    • Protector Multi-hazard Glove Boxes (discontinued)
    • Xpert and Rxpert Filtered Balance Systems
    • Xpert Filtered Balance Stations
    • Xpert Weigh Boxes
Specifications
Product Type Vibration Isolator
Color White, Gray
Material Stainless Steel
Includes Marble slab, Four isolator pad, Protective top cover