missing translation for 'onlineSavingsMsg'
Learn More

BAIAP2 Recombinant Protein, Abnova

Marca:  Abnova H00010458-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16195132

  • 295.00€ / 10 microgramos


Fecha estimada de envío
16-10-2020
Añadir a la cesta

Descripción

Descripción

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Especificaciones

Especificaciones

AAH32559
Solution
10458
BAIAP2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
BAP2|IRSP53
BAIAP2
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
82.06
8
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BAIAP2
Human
Yes
Videos
Documentos