Learn More
Honeywell™ BACOU TYLEX S3 Shoes
Honeywell™ BACOU TYLEX S3 Shoes feature oiled full grain leather uppers and stainless steel anti-puncture soles.
Marca:  Honeywell™ 6246120-48/7
Detalles adicionales : Peso : 2.38000kg
 Ver más versiones de este producto
Código de producto. 15826495
  • 76.50€ / 1 par
Fecha estimada de envío
14-11-2019
Añadir a la cesta
Descripción
Descripción
  • Oiled full grain leather upper provides great resistance and flexibility
  • Water resistant boot is perfectly suited to humid environments
  • Extra-wide pull-on lugs make the boots east to put on and take off
  • Few lateral seams make the upper more resistant in extreme environments
  • Midsole: Stainless steel: efficient technology covering the most extensive panel of risks. Puncture resistance: over 1100 N with a spike of 4.5 mm.
  • Upper: Oiled full grain leather. Extra-wide pull-on lugs. Few lateral seams
  • Toe Cap: 200J SPACIUM™ toe cap in injected polymer. Non-metallic: 50% lighter than steel. Extra-wide for greater comfort: suits all foot types. Non magnetic. Thermal insulator. Unalterable.
  • Insole: Dual density PU foam for intensive use. Removeable for easy care. Antibacterial for a longer life span and to prevent odour. Antistatic for optimal efficiency. Comfortable heel pads. Evacuation channels.
  • Outsole: Dual density polyurethane: PU 2D. Comfort layer of density: 0.5 (guarantees lightness and flexibility, ensures shock absorption and protects against changes in temperature). Outsole layer of density: 1 (resistance to abrasion, fuel oil and grease). High slip resistance due to well defined cleat design, wide, deep and self-cleaning. SRC certified: SRA: on ceramic plates : flat 0.45 (minimum standard: 0.32) heel 0.46 (minimum standard: 0.28) / SRB: on steel floor: flat 0.24 (minimum standard: 0.18) heel 0.24 (minimum standard: 0.13). Energy absorption capacity > 20 joules. Excellent resistance to abrasion: volume loss < 150 mm3 (with a load of 10 N = 1 kg). Extension before tearing: 650%. Optimised resistance to repeated flexing > 60,000 (standard >30,000). Resistant to acids and alkalis: limited to low concentrations.
Especificaciones
Especificaciones
EN ISO 20345:2011 standard, SRC certified
Leather
48
Brown, Black
Puncture resistance over 1100 N with a spike of 4.5 mm
Stainless Steel and Polyurethane