missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14018-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
BACH1 is a transcription factor that belongs to the cap′n′collar type of basic region leucine zipper factor family (CNC-bZip). The encoded protein contains broad complex, tramtrack, bric-a-brac/poxvirus and zinc finger (BTB/POZ) domains, which is atypical of CNC-bZip family members. These BTB/POZ domains facilitate protein-protein interactions and formation of homo and/or hetero oligomers. When this encoded protein forms a heterodimer with MafK, it functions as a repressor of Maf recognition element (MARE) and transcription is repressed. Multiple alternatively spliced transcript variants have been identified for this gene. Some exons of this gene overlap with some exons from the C21orf41 gene, which is transcribed in an opposite orientation to this gene, but which does not seem to encode a protein.Especificaciones
BACH1 | |
Polyclonal | |
Unconjugated | |
BACH1 | |
BACH 1, BACH1, BTB and CNC homolog 1, HA2303 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12013, 304127, 571 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O14867, P97302 | |
BACH1 | |
BACH1 Fusion Protein Ag5128 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |