missing translation for 'onlineSavingsMsg'
Learn More

Baboon Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NB300-664-0.05MG

Detalles adicionales : Peso : 0.00970kg

 Ver más versiones de este producto

Código de producto. 18215852

  • 417.92€ / 0.05 miligramo
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Baboon Polyclonal specifically detects Baboon in Drosophila samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Especificaciones

Especificaciones

Baboon
Polyclonal
Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500
Activin A receptor 45A, ATR 1, Atr 45A, Atr I, ATR1, Atr45A, AtrI, Bab, Babo, CG8224, G protein coupled receptor kinase 3, Gprk 3, Gprk3, l(2)k16912, RE55648p, STK E, STKE
Synthetic peptide: CNKNENMKRIFETDYVPHRLTS, corresponding to N terminal amino acids 200-221 of Drosophilia melanogaster Baboon. The peptide was amidated for conjugation.
Primary
Drosophila
IgG
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide
Rabbit
0.05 mg
The is unique to Baboon protein.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only