missing translation for 'onlineSavingsMsg'
Learn More

B7-H4 Antibody (188), Alexa Fluor™ 700, Novus Biologicals™

Rat Monoclonal Antibody

Marca:  Novus Biologicals NBP1-43266AF700

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213263

  • 650.09€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

B7-H4 Monoclonal specifically detects B7-H4 in Mouse samples. It is validated for Flow Cytometry.
Especificaciones

Especificaciones

B7-H4
Monoclonal
Alexa Fluor 700
B7h.5, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1VCTN1, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1
Rat
Affinity Purified
RUO
Primary
Mouse
IgG2b κ
Flow Cytometry
188
Flow Cytometry
VTCN1
The immunogen for this antibody was Purified B7H4.
0.1 mL
Cancer, Cell Cycle and Replication
79679
Store at 4C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only