Learn More
B220/CD45R Antibody (IVA103), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NB500-492AF488
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214250
  • 497.83€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
B220/CD45R Monoclonal specifically detects B220/CD45R in Bovine samples. It is validated for Flow Cytometry.
Especificaciones
Especificaciones
B220/CD45R
Monoclonal
Alexa Fluor 488
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Mouse
Affinity Purified
RUO
Primary
The antibody IVA103 reacts with CD45R antigen (bovine), a single chain type I transmembrane glycoprotein (member of the protein tyrosine phosphatase family), a restricted form of the CD45 family. CD45R is an essential regulator of T and B lymphocytes antigen receptor signaling and it is specifically expressed in hematopoietic cells.
Store at 4C in the dark.
Flow Cytometry
IVA103
Flow Cytometry
PTPRC
Bovine leukocytes.
0.1 mL
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
5788
Bovine
IgG1