Learn More
Brady™ B-489A Polyester Labels for BBP33 and i3300 Printers
Product Code. 15934550 Shop All Brady Products
Change view
Click to view available options
Height (Metric):
25.4 mm
50.8 mm
Thickness (Metric):
0.1 mm
0.13 mm
Vertical Repeat (Metric):
28.6 mm
54 mm
Width (Metric):
50.8 mm
76.2 mm
Unit Size:
Pack of 1500
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Thickness (Metric) Vertical Repeat (Metric) Width (Metric) Unit Size
15934550 50.8 mm 0.1 mm 54 mm 76.2 mm Pack of 500
15529294 25.4 mm 0.13 mm 28.6 mm 50.8 mm Pack of 1500
16278811 50.8 mm 0.13 mm 54 mm 76.2 mm Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15934550 Supplier Brady™ Supplier No. 142956
  • ONLINE EXCLUSIVE PRICE
    135.00€
    112.33€ / paquete de 500
    Ahorro 22.67€
    16% Descuento
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Polyester labels with permanent, rubber-based adhesive for use with B33 Series.
  • Permanent rubber adhesive
  • Abrasion resistant
  • For smooth, rough, or highly textured surfaces
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance UL Recognized, CSA Accepted, Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Permanent Rubber
Coating None
Color White
Finish Gloss
Format Die-cut
Material Polyester (B-483A)
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Asset Tracking, Barcode Labeling, Equipment Labeling, Electrical Labeling, General Identification
For Use With (Equipment) BBP™ 33, BradyPrinter i3300 and i5300 Industrial Label Printers
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
No. per Pack 500 Labels/Box
Quantity 500 Labels
Shape Rectangle
Temperature Range (English) -40°F to +248°F
Temperature Range (Metric) -40°C to +120°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.1 mm
Vertical Repeat (Metric) 54 mm
Dimensions (L x W) 50.80 x 76.2 mm
Length (English) 2 in.
Width (English) 3 in.
Width (Metric) 76.2 mm
Height (English) 2 in.
Height (Metric) 50.8 mm
Alternate Part Number B33-19-483