Learn More
Azlon™ Polypropylene Round Scoops
Sturdy scoops in rigid white polypropylene.
Marca: Azlon SWL204
Detalles adicionales : Peso : 0.17850kg
Descripción
- Suitable for food contact applications in accordance with U.S. FDA regulations
- Round design facilitates use when withdrawing substances from bags and sacks
Especificaciones
Round Scoop | |
250 mL | |
160 mm |
Polypropylene | |
White |