missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AXL, His Tag Recombinant Protein

The AXL Recombinant Human Protein is a chronic myelogenous leukemia-associated oncogene that is also associated with colon cancer and melanoma.

7890.00€

Especificaciones

Número de acceso NP_068713.2
Para utilizar con (aplicación) Kinase Assay
Peso molecular 48.3 kDa
Alias de gen JTK11, UFO
Familia Quinasa AXL Kinase Family
Ver más especificaciones

Productos 1
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
17198705
Ver documentos Detalles de la promoción
Invitrogen™
A31521
1 mg
7890.00€
1 miligramo
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 16-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

The AXL protein is a receptor tyrosine kinase that transduces signals from the extracellular matrix into the cytoplasm by binding growth factors like vitamin K-dependent protein growth-arrest-specific gene 6, and is involved in the stimulation of cell proliferation.

The purified recombinant protein can be used in a number of different applications, but it has been specifically validated for use with both the LanthaScreen™ and Z'-LYTE™ assay technologies to identify novel kinase inhibitors.

  • Validated for use with the LanthaScreen and Z'-LYTE assay technology
  • Available in multiple pack sizes for convenient use
Especificaciones

Especificaciones

NP_068713.2
48.3 kDa
AXL Kinase Family
Human
His Tag
Dry Ice
Kinases & Inhibitors
Liquid
Kinase Assay
JTK11, UFO
473 - end
Recombinant
Storage Buffer: 50 mM sodium phosphate, pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, 25% glycerol

Store at -80°C. Upon first use, aliquot and store at -80°C to avoid multiple freeze/thaws.
Baculovirus
Recombinant, Active, Cytoplasmic, Truncated
Signal Transduction
Videos
SDS
Documentos

Documentos

Product Certifications