Learn More
Axl Monoclonal Antibody (MAXL8DS), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
en la oferta
Especificaciones
Antígeno | Axl |
---|---|
Clon | MAXL8DS |
Concentración | 0.2 mg/mL |
Aplicaciones | Flow Cytometry |
Clasificación | Monoclonal |
Descripción
Description: This MAXL8DS monoclonal antibody recognizes mouse Axl, a member of the TAM family of tyrosine kinase receptors that also includes MerTK and Tyro3. Within hematopoietic compartment the highest expression of Axl can be observed on dendritic cells and macrophages. Stimulation with TLR ligands has been shown to additionally upregulate this expression. This MAXL8DS antibody will work in flow cytometry on both native and fixed/permeabilized cells. Applications Reported: This MAXL8DS antibody has been reported for use in flow cytometric analysis. Applications Tested: This MAXL8DS antibody has been tested by flow cytometric analysis of mouse bone marrow cells cultured in the presence of GM-CSF. This may be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No.
Axl Monoclonal antibody specifically detects Axl in Mouse samples. It is validated for Flow Cytometry.Especificaciones
Axl | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
Adhesion-related kinase; AI323647; ARK; AXL; AXL oncogene; AXL receptor tyrosine kinase; AXL transforming sequence/gene; AZF1; JTK11; oncogene AXL; sAXL; soluble AXL; Tyro7; Tyrosine-protein kinase receptor UFO; Ufo; ufo oncogene homolog | |
AXL | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
MAXL8DS | |
Flow Cytometry | |
PE-Cyanine7 | |
Rat | |
Mouse | |
Q00993 | |
26362 | |
C57BL/6 mouse Axl-human IgG Fc fusion protein | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
AXL |