missing translation for 'onlineSavingsMsg'
Learn More

AWAT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 25119-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16813665

  • 319.00€ / 150 microlitros

Envío estimado: 22-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes an enzyme belonging to the diacylglycerol acyltransferase family. This enzyme produces wax esters by the esterification of long chain (or wax) alcohols with acyl-CoA-derived fatty acids. It functions in lipid metabolism in the skin, mostly in undifferentiated peripheral sebocytes. This enzyme may also have acyl-CoA:retinol acyltransferase activities, where it catalyzes the synthesis of diacylglycerols and retinyl esters.
Especificaciones

Especificaciones

AWAT2
0.21 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6E1M8, Q6E213
AWAT2, DC4, DGAT2L4, hDC4, hWS, MFAT, Wax synthase, WS
Rabbit
IgG
150 μL
-20°C
Primary
158835, 245532, 302425
Western Blot
Unconjugated
AWAT2
Liquid
Awat2
AWAT2 Fusion Protein Ag18936
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados