Learn More
Invitrogen™ Avidin, FITC conjugate
Product Code. 11554137
Change view
Click to view available options
Packaging:
1 Each
Unit Size:
5mg
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Unit Size
11554137 1 Each 5mg
10009232 - Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11554137 Supplier Invitrogen™ Supplier No. A821
 Restricted Item
En stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Avidin, FITC conjugate
Our avidin, fluorescein conjugate has excitation and emission maxima ∼494/518. Avidin is a glycosylated biotin-binding protein that has a basic isoelectric point and is widely used to detect biotinylated probes.
TRUSTED_SUSTAINABILITY
Specifications
Conjugate FITC (Fluorescein)
Quantity 5 mg
Packaging 1 Each
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
Product Type Avidin
For Research Use Only. Not for use in diagnostic procedures.