Learn More
Invitrogen™ Avidin, Egg White
Product Code. 11564137
Change view
Click to view available options
Quantity:
5 mg
100 mg
Unit Size:
100mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11564137 100 mg 100mg
11554037 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11564137 Supplier Invitrogen™ Supplier No. A887
 Restricted Item
Fecha estimada de envoi: 04-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Avidin, egg white, 5 mg
Avidin from egg white is a glycosylated biotin-binding protein that has a basic isoelectric point and is widely used to detect biotinylated probes.
TRUSTED_SUSTAINABILITY
Specifications
Conjugate Unconjugated
Quantity 100 mg
Content And Storage Store in freezer (-5 to -30°C).
Shipping Condition Room Temperature
For Research Use Only. Not for use in diagnostic procedures.