Learn More
- Home
- Products
- Chromatography Syringes
- Autosampler Syringes
- Trajan™ Autosampler Syringes with Removable Needle for CTC Analytics CombiPAL and GC PAL
Trajan™ Autosampler Syringes with Removable Needle for CTC Analytics CombiPAL and GC PAL
Product Code. 13189015
Change view
Click to view available options
Needle Gauge:
23 G
26 G
Diameter (English) Outer Needle:
0.01 in.
0.018 in.
0.02 in.
Length (English) Needle:
1.9 in.
2.2 in.
3.3 in.
Model:
10F-RTC/RSH-5.7/0.47C
10F-RTC/RSH-5.7/0.63C
10F-RTC/RSH-8.5/0.47C
10F-RTC/RSH-8.5/0.63C
10F-RTC/RSH-GT-5.7/0.47C
10F-RTC/RSH-GT-5.7/0.63C
10F-RTC/RSH-GT-8.5/0.47C
10F-RTC/RSH-GT-8.5/0.63C
25F-RTC/RSH-GT-5.7/0.47C
25F-RTC/RSH-GT-5.7/0.63C
25F-RTC/RSH-GT-8.5/0.63C
25FRTC/RSH-GT-8.5/0.47C
25R-C/T-GT-0.47
50F-RTC/RSH-GT-5.7/0.47C
50F-RTC/RSH-GT-5.7/0.63C
50F-RTC/RSH-GT-8.5/0.47C
50F-RTC/RSH-GT-8.5/0.63C
5F-RTC/RSH-5.7/0.47C
5F-RTC/RSH-5.7/0.63C
5F-RTC/RSH-8.5/0.63C
Unit Size:
Each
20 product options available for selection
| Product Code. | Needle Gauge | Diameter (English) Outer Needle | Length (English) Needle | Model | Unit Size |
|---|---|---|---|---|---|
| 13189015 | 26 G | 0.01 in. | 2.2 in. | 10F-RTC/RSH-5.7/0.47C | Each |
| 13186105 | 23 G | 0.02 in. | 2.2 in. | 5F-RTC/RSH-5.7/0.63C | Each |
| 13199195 | 26 G | 0.01 in. | 2.2 in. | 10F-RTC/RSH-GT-5.7/0.47C | Each |
| 13179015 | 23 G | 0.02 in. | 3.3 in. | 10F-RTC/RSH-8.5/0.63C | Each |
| 13109205 | 26 G | 0.01 in. | 3.3 in. | 10F-RTC/RSH-GT-8.5/0.47C | Each |
| 13109215 | 23 G | 0.02 in. | 3.3 in. | 50F-RTC/RSH-GT-8.5/0.63C | Each |
| 13106115 | 26 G | 0.01 in. | 2.2 in. | 5F-RTC/RSH-5.7/0.47C | Each |
| 13169015 | 23 G | 0.02 in. | 2.2 in. | 10F-RTC/RSH-5.7/0.63C | Each |
| 13196105 | 23 G | 0.02 in. | 3.3 in. | 5F-RTC/RSH-8.5/0.63C | Each |
| 13129215 | 26 G | 0.01 in. | 3.3 in. | 50F-RTC/RSH-GT-8.5/0.47C | Each |
| 13199205 | 23 G | 0.02 in. | 2.2 in. | 50F-RTC/RSH-GT-5.7/0.63C | Each |
| 13179195 | 23 G | 0.02 in. | 2.2 in. | 10F-RTC/RSH-GT-5.7/0.63C | Each |
| 13189205 | 26 G | 0.01 in. | 3.3 in. | 25FRTC/RSH-GT-8.5/0.47C | Each |
| 13179205 | 26 G | 0.01 in. | 2.2 in. | 25F-RTC/RSH-GT-5.7/0.47C | Each |
| 13119215 | 26 G | 0.01 in. | 2.2 in. | 50F-RTC/RSH-GT-5.7/0.47C | Each |
| 13189195 | 23 G | 0.02 in. | 3.3 in. | 10F-RTC/RSH-GT-8.5/0.63C | Each |
| 13159205 | 23 G | 0.02 in. | 2.2 in. | 25F-RTC/RSH-GT-5.7/0.63C | Each |
| 13169205 | 23 G | 0.02 in. | 3.3 in. | 25F-RTC/RSH-GT-8.5/0.63C | Each |
| 13199015 | 26 G | 0.01 in. | 3.3 in. | 10F-RTC/RSH-8.5/0.47C | Each |
| 12977506 | 26 G | 0.018 in. | 1.9 in. | 25R-C/T-GT-0.47 | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
This item is not returnable.
View return policy
Product Code.
13189015
Supplier
Trajan™
Supplier No.
002865
-
84.30€
64.25€ / paquete de 1
Ahorro 20.05€
23% Descuento
Fecha estimada de envoi: 05-01-2027
Inicie sesión para ver el stock
Inicie sesión para ver el stock
Add to basket
Add to basket
This item is not returnable.
View return policy
Description
- Borosilicate glass barrel for strength and inertness.
- Stainless steel needles for inertness, strength and corrosion resistance.
- Backing behind the scale aids quick and accurate reading of the syringe scale.
- Metal back flange provides stability and resists fracture.
- Metal needle cap on syringe barrel protects and improves syringe lifetime.
- Syringes are individually tested for quality ensuring reliability and conformance.
- Unique packaging design eliminates possible contamination in the syringe.
Specifications
Specifications
| Needle Gauge | 26 G |
| Diameter (English) Outer Needle | 0.01 in. |
| Diameter (Metric) Outer Needle | 0.47 mm |
| For Use With (Application) | Gas chromatography |
| Length (English) Needle | 2.2 in. |
| Product Type | Autosampler Syringe |
| Model | 10F-RTC/RSH-5.7/0.47C |
| Quantity | Ea. |
| Volume (Metric) | 10 μL |
| Needle Point Style | Cone |
| For Use With (Equipment) | Thermo RSH Autosampler |
| Connection Type | Fixed |
| Show More Show Less |