Learn More
Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with MP20 and MP200 Dispense Heads
Product Code. 11791223 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Volume (Metric):
20 μL
130 μL
200 μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
Unit Size:
Pack of 4800
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Filtered Sterility Fit for Purpose Unit Size
11791223 200 μL Non-filtered Non-sterile Certified Quality Pack of 4800
11781263 20 μL Filtered Sterile Sterile and PCR Ready Pack of 4800
11771263 20 μL Non-filtered Sterile Sterile Pack of 4800
11781223 20 μL Non-filtered Non-sterile Certified Quality Pack of 4800
11741253 130 μL Filtered Sterile Sterile and PCR Ready Pack of 4800
11751253 200 μL Non-filtered Sterile Sterile Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11791223 Supplier Thermo Scientific™ Supplier No. 917262
  • ONLINE EXCLUSIVE PRICE
    480.00€
    445.00€ / paquete de 4800
    Ahorro 35.00€
    7% Descuento
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well plates
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Format Rack
Tip Style Extended Length
For Use With (Equipment) Beckman Coulter Biomek 1000/2000
Product Line Beckman
Fit for Purpose Certified Quality