Learn More
Deltalab™ Autoclave Resistant Bags
Product Code. 12064363
Change view
Click to view available options
Dimensions (L x W):
30 x 60cm
40 x 75cm
60 x 75cm
75 x 50cm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
12064363 40 x 75cm Each
12004353 75 x 50cm Each
12054363 30 x 60cm Each
12074363 60 x 75cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12064363 Supplier Deltalab™ Supplier No. 200311
En stock
Add to basket
Edge
Add to basket
Wrap autoclaveable items in Deltalab™Autoclave Resistant Bags when working with laboratory items requiring sterilization.
  • Printed with biohazard graphic symbol and use instructions in five languages
  • ags placed in autoclave at 121ºC. should not be sealed hermetically
  • Minimum order quantity: 50 pieces
Specifications
Autoclavable Autoclavable
Quantity 300 Pack
Dimensions (L x W) 40 x 75cm