missing translation for 'onlineSavingsMsg'
Learn More

Aurora A Antibody (1F8) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-80587

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214346

  • 451.39€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

Aurora A Monoclonal specifically detects Aurora A in Human, Rat, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
Especificaciones

Especificaciones

Aurora A
Monoclonal
1 mg/mL
Western Blot : 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA : 1:10000, Immunocytochemistry/Immunofluorescence : 1:200-1:1000, CyTOF-ready
AIKAurA, ARK-1, ARK1EC 2.7.11.1, AURA, aurora kinase AhARK1, aurora/IPL1-like kinase, Aurora/IPL1-related kinase 1, Aurora-related kinase 1, Breast tumor-amplified kinase, breast-tumor-amplified kinase, BTAKAURORA2, EC 2.7.11, IPL1-related kinase, serine/threonine kinase 6, serine/threonine protein kinase 15, Serine/threonine-protein kinase 15, serine/threonine-protein kinase 6, Serine/threonine-protein kinase aurora-A, STK15serine/threonine kinase 15, STK6MGC34538, STK7
Mouse
Ammonium sulfate precipitation
RUO
Primary
Human, Rat, Primate
Purified
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF
1F8
Unconjugated
PBS
AURKA
Purified recombinant fragment of human Aurora A expressed in E. coli. [Uniprot: O14965]
0.1 mL
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, MAP Kinase Signaling, Mitotic Regulators, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers, Tumor Suppressors
6790
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales