Learn More
Invitrogen™ Attune™ NxT and CytPix Maintenance Kit
Product Code. 17174729
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
Product Code. Quantity Unit Size
17174729 1 kit Each
1 options
This item is not returnable. View return policy
Product Code. 17174729 Supplier Invitrogen™ Supplier No. A43038
 Restricted Item
Fecha estimada de envoi: 03-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The Attune NxT Maintenance Kit will help you avoid down-time and ensure optimal operation of your Attune NxT Acoustic Focusing Cytometer hardware.
This convenient kit includes sufficient supplies to perform all recommended preventative maintenance procedures for 12 months. These procedures are: replacement of the syringe (bi-yearly), SIP tube (yearly), and instrument bottles (yearly), as well as performance of system decontamination (quarterly). The kit includes 1 waste bottle, 1 focusing fluid tank, 12 focusing fluid filters, 2 syringes, 1 SIP tube, and 1 debubble solution.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Attune
Content And Storage • 1 waste bottle
• 1 focusing fluid bottle
• 12 focusing fluid filters
• 2 syringes
• 1 SIP
• 1 debubble solution
Store at room temperature.
Quantity 1 kit
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Block Format Non-Interchangeable
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Maintenance Kit
For Research Use Only. Not for use in diagnostic procedures.