Learn More
Applied Biosystems™ Attune™ Flow Cytometer Syringe
Product Code. 17246493 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 mL
10 mL
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17246493 10 mL 10mL
15830413 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17246493 Supplier Applied Biosystems™ Supplier No. 4476090
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
The sample syringe is used with the fluidics system of the Attune Flow Cytometer to move sample to the flow cell.
  • The sample syringe is used with the fluidics system of the Attune Flow Cytometer to move sample to the flow cell.
  • Replacement of this syringe is recommended as part of instrument maintenance.
TRUSTED_SUSTAINABILITY
Specifications
Product Line Attune
Content And Storage Store at room temperature
Quantity 10 mL
Shipping Condition Room Temperature
For Use With (Equipment) Attune Flow Cytometer
Product Type Syringe
For Research Use Only. Not for use in diagnostic procedures.