Learn More
Novus Biologicals™ ATPB Overexpression Lysate
Marca:  Novus Biologicals™ NBL1-07817
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213325
  • 685.73€ / 0.10 miligramo
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
The ATPB Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Especificaciones
Especificaciones
Western Blot
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
The lysate was created in HEK293T cells, using plasmid ID RC21638 and based on accession number NM_1686. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide (ATP5B), nuclear gene encoding mitochondrial protein, mRNA.
Native