missing translation for 'onlineSavingsMsg'
Learn More

ATP6V1H Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP1-32980

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215810

  • 535.62€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

ATP6V1H Polyclonal specifically detects ATP6V1H in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

ATP6V1H
Polyclonal
Unconjugated
PBS, 20% Glycerol (pH7) with 0.025% Proclin 300
ATP6V1H
Recombinant protein encompassing a sequence within the center region of human ATP6V1H. The exact sequence is proprietary.
0.1 mL
Primary
Tested in human. Not yet tested in any other species.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.56mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000
ATPase, H+ transporting, lysosomal 50/57kD, V1 subunit H, ATPase, H+ transporting, lysosomal 50/57kDa, V1 subunit H, CGI-11, MSTP042, NBP1, Nef-binding protein 1, Protein VMA13 homolog, SFD, SFDalpha, SFDbeta, vacuolar ATP synthase subunit H, vacuolar ATPase subunit H, vacuolar proton pump H subunit, Vacuolar proton pump subunit H, Vacuolar proton pump subunit SFD, V-ATPase 50/57 kDa subunits, V-ATPase H subunit, V-ATPase subunit H, VMA13, V-type proton ATPase subunit H
Rabbit
Affinity Purified
RUO
51606
Human, Mouse
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only