missing translation for 'onlineSavingsMsg'
Learn More

ATP1B4 Antibody (108), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-90158R

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214128

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

ATP1B4 Monoclonal specifically detects ATP1B4 in Human samples. It is validated for Western Blot, ELISA.
Especificaciones

Especificaciones

ATP1B4
Monoclonal
DyLight 550
50mM Sodium Borate
Rabbit
Protein A purified
RUO
23439
Store at 4°C in the dark.
IgG
Western Blot, ELISA
108
Western Blot, ELISA
ATPase, (Na+)/K+ transporting, beta 4 polypeptide, ATPase, Na+/K+ transporting, beta 4 polypeptide, K-ATPase beta-m subunit, K-ATPase subunit beta-m, protein ATP1B4, X, X/potassium-transporting ATPase subunit beta-m
This antibody was obtained from a rabbit immunized with purified, recombinant Human ATP1B4 (mature form, NP_036201.1; Tyr 132-Thr 357).
0.1 mL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales