missing translation for 'onlineSavingsMsg'
Learn More

ATG2B Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Novus Biologicals NBP2-81706AF488

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214358

  • 577.74€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

ATG2B Polyclonal antibody specifically detects ATG2B in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Especificaciones

Especificaciones

ATG2B
Polyclonal
50mM Sodium Borate
Rabbit
Peptide affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA, Immunofluorescence
Alexa Fluor 488
ATG2 autophagy related 2 homolog B (S. cerevisiae), C14orf103, FLJ10242
ATG2B antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human ATG2B. The immunogen is located within amino acids 450 - 500 of ATG2B. Amino Acid Squence: EFLDHHKEQPVRGSTFPSN
0.1 mL
Macroautophagy
55102
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales