missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG16L2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24322-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
May play a role in autophagy By similarity. Ota T., Nat. Genet. 36:40-45(2004). Mural R. J., Submitted (JUL-2005) to the EMBL/GenBank/DDBJ databases. Bechtel S., BMC Genomics 8:399-399(2007).Especificaciones
ATG16L2 | |
Polyclonal | |
Unconjugated | |
ATG16L2 | |
APG16 like 2, ATG16L2, Autophagy related protein 16 2, WDR80 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
73683, 89849 | |
-20°C | |
Liquid |
Immunofluorescence, Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6KAU8, Q8NAA4 | |
Atg16l2 | |
ATG16L2 Fusion Protein Ag19399 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |