missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASUN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19892-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Crucial regulator of the mitotic cell cycle and development.Especificaciones
ASUN | |
Polyclonal | |
Unconjugated | |
INTS13 | |
ASUN, C12orf11, GCT1, NET48, NY SAR 95, Sarcoma antigen NY SAR 95 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55726, 690728, 71177 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8QZV7, Q9NVM9 | |
Ints13 | |
ASUN Fusion Protein Ag13727 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |