Learn More
Thermo Scientific™ SoftFit-L™ Filtered Pipette Tips in Hinged Racks
Product Code. 11585454
Change view
Click to view available options
Volume (Metric):
20 μL
200 μL
300 μL
1000 μL
1200 μL
Unit Size:
Pack of 768
Pack of 960
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11585454 200 μL Pack of 960
12994037 1000 μL Pack of 768
11565454 20 μL Pack of 960
11505464 300 μL Pack of 960
11373393 1200 μL Pack of 768
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11585454 Supplier Thermo Scientific™ Supplier No. 2769HR
  • ONLINE EXCLUSIVE PRICE
    216.00€
    133.80€ / paquete de 960
    Ahorro 82.20€
    38% Descuento
View all promotions
En stock
Add to basket
Edge
Add to basket
Bulk Quote Request
Pipette samples with the accuracy and precision you expect from your pipette, every time with these pipette tips. For use with Rainin™ LTS pipettes.
Pipette samples with the accuracy and precision you expect from your pipette, every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
  • Hinged rack provides easy one-hand access to tips
  • Color-coded inserts allow quick identification of tip volume
TRUSTED_SUSTAINABILITY
Specifications
Description 200 μL, Filtered, Sterile, Hinged Rack
Volume (Metric) 200 μL
Filtered Filtered
Sterility Sterile
Color Natural
Model 200L
Length (English) 1.97 in.
Length (Metric) 5.00cm
Clean Claims DNase, RNase, DNA, ATP, and Endotoxin Free
Format Hinged Rack
Tip Style Universal
Color Code Green
For Use With (Equipment) Rainin LTS LiteTouch
Product Line ART SoftFit L
Fit for Purpose Sterile & PCR Ready