Learn More
Arsenic(V) oxide, 99.9+%, (trace metal basis), ACROS Organics™
Product Code. 10394492 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
Product Code.
10394492 25g; Glass bottle
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10394492 Supplier Thermo Scientific Acros Supplier No. 366310250
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
For ultra trace analysis
TRUSTED_SUSTAINABILITY
Specifications
CAS 1303-28-2
Color White
Physical Form Chunks and/or Powder
Quantity 25 g
Assay Percent Range 99.9+%
Molecular Formula As2O5
Linear Formula As2O5
MDL Number MFCD00003434
Synonym arsenic pentoxide, arsenic v oxide, arsenic anhydride, arsenic v pentoxide, arsenic pentaoxide, arsenic oxide, diarsenic pentoxide, diarsenic pentaoxide, arsenic 5+ oxide, arsenic acid anhydride
InChI Key COHDHYZHOPQOFD-UHFFFAOYSA-N
SMILES O=[As](=O)O[As](=O)=O
Molecular Weight (g/mol) 229.838
PubChem CID 14771
Formula Weight 229.84
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Arsenic(V) oxide, 99.9+%