missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Armenian Hamster IgG Isotype Control (eBio299Arm), Super Bright™ 780, eBioscience™, Invitrogen™
Armenian Hamster Isotype Control
en la oferta
Especificaciones
Antígeno | Armenian Hamster IgG |
---|---|
Clon | eBio299Arm |
Concentración | 0.2 mg/mL |
Conjugado | Super Bright 780 |
Formulario | Liquid |
Descripción
Armenian Hamster IgG Control antibody specifically detects Armenian Hamster IgG in Not Applicable samples. It is validated for Control, Flow Cytometry.Especificaciones
Armenian Hamster IgG | |
0.2 mg/mL | |
Liquid | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Isotype Control (Primary) |
eBio299Arm | |
Super Bright 780 | |
RUO | |
IgG | |
Affinity chromatography |