missing translation for 'onlineSavingsMsg'
Learn More

Armenian Hamster IgG, Super Bright 702, eBioscience™

Armenian Hamster Isotype Control

Marca:  Invitrogen 67-4888-80

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923764

  • 134.00€ / 25 microgramos


Fecha estimada de envío
20-08-2020
Añadir a la cesta

Descripción

Descripción

Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100μL of cell sample + 100μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405nm; Emission: 702nm; Laser: Violet Laser

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody and quote;s species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.

Especificaciones

Especificaciones

Armenian Hamster IgG
Control
Super Bright 702
Liquid
IgG
25μg
4°C, store in dark, DO NOT FREEZE!
Not Applicable
Control, Flow Cytometry
0.2mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
Armenian Hamster
Affinity chromatography
RUO
Isotype Control
Videos
Documentos