missing translation for 'onlineSavingsMsg'
Learn More

Armenian Hamster IgG, Super Bright 436, eBioscience™

Armenian Hamster Isotype Control

Marca:  Invitrogen 62-4888-82

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933424

  • 257.00€ / 100 microgramos


Fecha de envío estimada
del proveedor 26-11-2019
Añadir a la cesta

Descripción

Descripción

The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Especificaciones

Especificaciones

Armenian Hamster IgG
Control
0.2mg/mL
PBS with BSA and 0.09% sodium azide; pH 7.2
Armenian Hamster
Affinity chromatography
RUO
Isotype Control
Control, Flow Cytometry
eBio299Arm
Super Bright 436
Liquid
IgG
100μg
4°C, store in dark, DO NOT FREEZE
Not Applicable
Videos
Documentos