Learn More
Sartorius arium™ Pre-Treatment Cartridge
Product Code. 15648938 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
arium™ systems
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15648938 arium™ systems Each
1 options
Product Code. 15648938 Supplier Sartorius Supplier No. H2OCPFAD1
  • ONLINE EXCLUSIVE PRICE
    489.00€
    417.00€ / paquete de 1
    Ahorro 72.00€
    14% Descuento
View all promotions
 Restricted Item
En stock
Add to basket
Edge
Add to basket
For the filtration of water in lab water systems.
  • Accessory item
  • For use with arium lab water systems
  • Aids in applications such as HPLC, ion chromatography, TOC analysis, PCR, electrophoresis, photometry and many more
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Elix, RiOs
Material Polypropylene
Purification Method Activated Carbon
Resin Type Activated Carbon
For Use With (Application) Lab water filtration
For Use With (Equipment) arium™ systems
Type Cartridge