missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGDIB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16122-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.Especificaciones
ARHGDIB | |
Polyclonal | |
Unconjugated | |
ARHGDIB | |
ARHGDIB, D4, GDIA2, GDID4, Ly GDI, LYGDI, RAP1GN1, Rho GDI 2, Rho GDI beta, RhoGDI2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11857, 362456, 397 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P52566, Q61599 | |
Arhgdib | |
ARHGDIB Fusion Protein Ag9078 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |