missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ARHGDIB (Human) Recombinant Protein
Human ARHGDIB full-length ORF ( AAH09200, 1 a.a. - 201 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00000397-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MTEKAPEPHVEEDDDDELDSKLNYKPPPQKSLKELQEMDKDDESLIKYKKTLLGDGPVVTDPKAPNVVVTRLTLVCESAPGPITMDLTGDLEALKKETIVLKEGSEYRVKIHFKVNRDIVSGLKYVQHTYRTGVKVDKATFMVGSYGPRPEEYEFLTPVEEAPKGMLARGTYHNKSFFTDDDKQDHLSWEWNLSIKKEWTE
Especificaciones
AAH09200 | |
Rho GDP dissociation inhibitor (GDI) beta | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARHGDIB | |
GST |
397 | |
Wheat germ expression system | |
25 μg | |
D4, GDIA2, GDID4, LYGDI, Ly-GDI, RAP1GN1, RhoGDI2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |