missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ARHGDIA (Human) Recombinant Protein
Human ARHGDIA full-length ORF ( AAH05851, 1 a.a. - 204 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00000396-P02.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MAEQEPTAEQLAQIAAENEEDEHSVNYKPPAQKSIQEIQELDKDDESLRKYKEALLGRVAVSADPNVPNVVVTGLTLVCSSAPGPLELDLTGDLESFKKQSFVLKEGVEYRIKISFRVNREIVSGMKYIQHTYRKGVKIDKTDYMVGSYGPRAEEYEFLTPVEEAPKGMLARGSYSIKSRFTDDDKTDHLSWEWNLTIKKDWKD
Especificaciones
AAH05851 | |
Rho GDP dissociation inhibitor (GDI) alpha | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARHGDIA | |
GST |
396 | |
Wheat germ expression system | |
10 μg | |
GDIA1, MGC117248, RHOGDI, RHOGDI-1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |