missing translation for 'onlineSavingsMsg'
Learn More

ARHGAP18 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Novus Biologicals NBP2-81704B

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213987

  • 577.74€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

ARHGAP18 Polyclonal antibody specifically detects ARHGAP18 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Especificaciones

Especificaciones

ARHGAP18
Polyclonal
PBS
Rabbit
Peptide affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA, Immunofluorescence
Biotin
bA307O14.2, MacGAP, MGC126757, Rho GTPase activating protein 18, rho-type GTPase-activating protein 18
ARHGAP18 antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ARHGAP18. The immunogen is located within the last 50 amino acids of ARHGAP18. Amino Acid Squence: DTYMKDLYQLNPNAE
0.1 mL
Signal Transduction
93663
Store at 4°C in the dark.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales