Learn More
Arginase 1/ARG1/liver Arginase Antibody (034), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca:  Bio-Techne NBP2-89915AF405
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213539
  • 607.98€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
Arginase 1/ARG1/liver Arginase Monoclonal specifically detects Arginase 1/ARG1/liver Arginase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
Arginase 1/ARG1/liver Arginase
Monoclonal
Alexa Fluor 405
arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, Liver-type arginase, Type I arginase
This antibody was obtained from a rabbit immunized with purified, recombinant Human Arginase 1/ARG1/liver Arginase (P05089-1; Met 1-Lys 322).
0.1 mL
Cancer, Cellular Markers, Chromatin Research, Lipid and Metabolism, Myeloid derived Suppressor Cell
383
Store at 4°C in the dark.
IgG
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
34
50 mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Human
Purified