missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARFRP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17712-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a membrane-associated GTP-ase which localizes to the plasma membrane and is related to the ADP-ribosylation factor (ARF) and ARF-like (ARL) proteins. This gene plays a role in membrane trafficking between the trans-Golgi network and endosomes. Alternatively spliced transcript variants encoding different isoforms have been identified.Especificaciones
ARFRP1 | |
Polyclonal | |
Unconjugated | |
ARFRP1 | |
ARF related protein 1, ARFRP1, ARL18, ARP, Arp1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10139, 117051, 76688 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13795, Q63055, Q8BXL7 | |
Arfrp1 | |
ARFRP1 Fusion Protein Ag12036 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |