missing translation for 'onlineSavingsMsg'
Learn More

ARF1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10790-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817413

  • 109.00€ / 20 microlitros

Envío estimado: 29-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This antibody can bind ARFs for the close sequences.

ADP-ribosylation factor 1 (ARF1) is a member of the human ARF gene family. The family members encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking as activators of phospholipase D. The gene products, including 6 ARF proteins and 11 ARF-like proteins, constitute a family of the RAS superfamily. The ARF proteins are categorized as class I (ARF1, ARF2 and ARF3), class II (ARF4 and ARF5) and class III (ARF6), and members of each class share a common gene organization. The ARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
Especificaciones

Especificaciones

ARF1
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P84077, P84078, P84079
ADP ribosylation factor 1, ARF1
Rabbit
IgG
20 μL
-20°C
Primary
11840, 375, 64310
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ARF1
Liquid
ARF1
ARF1 Fusion Protein Ag1244
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Non-human Primate, Mouse, Rat, Human
Videos
SDS
Documentos
Certificados